Blot Development Folders, Azure Biosystems
10147-346
Product available on GSA Advantage®
- Description:Blot Development Folders
- Cat. no.:10147-346
- Packaging:Pack of 100
- Supplier No.:AC2126
These transparent plastic folders hold your blots during handling and imaging
During handling, the folders protect your blots from dust and from scratches from forceps.
During imaging, the folders provide a wrinkle-free alternative to plastic wrap.