Radiance Q Chemiluminescent Substrate, Azure Biosystems
Catalogue # 10147-296
Fournisseur: Azure Biosystems
Product available on GSA Advantage®
Numéro CAS:
Spécifications
- Description:HRP Substrate for CCD Kit
- Size:150 mL
- Cat. no.:10147-296
- Supplier No.:AC2101
Spécifications
À propos de cet article
Radiance Q chemiluminescent substrate sets the bar for sensitivity and quantitative ability
- Substrate for 1500 cm2 Membrane
Specially developed for CCD imaging, Radiance Q produces a strong, long-lasting signal with extremely low background, perfect for detecting low abundance proteins.
Since it does not exhibit substrate depletion at high protein loads, Radiance Q provides the largest dynamic range of any chemiluminescent substrate for the most quantitative chemiluminescent western experiments.