Cy™ Amidites
Fournisseur: Cytiva
Certificats
À propos de cet article
Cy™ fluorescent amidites for incorporation of Cy3, Cy3.5, Cy5, Cy5.5 fluorescent dyes at any position in an oligonucleotide during synthesis.
- For incorporation of Cy fluorescent dyes at any position in an oligonucleotide during synthesis
- Linkage of Cy dyes allows labeled DNA to hybridize to other DNA
Cy5 amidite can be used to prepare sequencing primers for use on ALF express.
Delivery: Larger pack sizes and custom packaging available upon request.
Spécifications
- Storage conditions:Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, away from incompatible materials. Keep container tightly closed and sealed until ready for use
- Solubility:Soluble in cold water and hot water
- Stability:Stable
- Physical form:Solid