Phosphate Buffered Saline (PBS), Quality Biological
Supplier: QUALITY BIOLOGICAL, INC.
Certificates
About this item
PBS (phosphate buffered saline) is a balanced salt solution used for a variety biological and cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents.
- Endotoxin and Performance tested
PBS is also useful in immunological applications, such as Western Blotting.
Certifications: Manufactured under a cGMP compliant quality management system in an ISO 9001:2015 certified facility.
Caution: Research use only.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile filtered
- Shipping conditions:Ambient temperature
- Test description:Endotoxin and mycoplasma tested
- Shelf life:24 months
- Osmolality:300 ±5% mOsm/Kg H₂O
- Shipping temperature:Ambient