KIMBLE® Outer Joint Hose Inlet Adapter, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Ideal for use as a vent or for the introduction of pressure/vacuum.
- [ST] joint at one end
- Vertical hose connection at other end
- Manufactured from borosilicate glass
Adapter features a Standard Taper joint at one end and vertical hose connection at the other.
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together