KIMBLE® BEVEL-SEAL™ Thermometer Inlet Adapters with Hose Connection, DWK Life Sciences
Designed for use with plain thermometers, pipets, syringes, or other small items with an O.D. from 2mm to 11mm.
- Vacuum-tight seal
- With hose connection
- One FKM O-Ring and one cap included
Equipped with a hose connection for vacuum or the introduction of gases
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
A vacuum-tight seal is created with the open-top compression cap and FKM O-Ring to allow adjustable immersion of thermometers, etc. Cap is suitable for use to 150°C (302°F). To use as a septum port, replace the O-Ring with a PTFE-lined septum. Supplied with one FKM O-Ring and one cap.
Frequently Bought Together