Plastic Bumper, KIMBLE®, DWK Life Sciences
Bumper, size 4, fits 28.5 - 31.0 mm (100 ml) cylinder OD
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 20100-4
Jump to:
Product Details & Documents
When placed around a cylinder near the top, this sturdy split ring protects against breakage if the cylinder is upset
Made of yellow low density polyethylene. Position of ring can be adjusted to avoid contact with the top of a receiver when liquid is poured from the cylinder. Should a ring become loose after prolonged use, soak it briefly in hot water (90 °C) to return it to its original shape.
Specifications
- Désignation:Bumper, size 4, fits 28.5 - 31.0 mm (100 ml) cylinder OD