KimCote® Cylinders with Reverse Graduations, Class A, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
KimCote® safety coating reduces the hazards of shattered glass
Designed from ASTM Specification E1272, Style I, Class A requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Each cylinder is marked with “TD” which indicates the cylinder is calibrated to deliver, and a white, reverse, single metric scale. The hexagonal base is flat ground for stability and the SAFE-GARD® bumper is supplied.
Specifications
- Catalog No:
- 89097-390
- 89097-388
- 89097-386
- 89097-384
- 89097-382
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall