KIMAX® Pickel Extraction Flask, Kimble Chase
Catalog # 29140-607
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Ext.Ø:45 mm
- Height:133 mm
- Cat. no.:29140-607
- Supplier No.:26850-99
Specifications
About this item
KIMAX® flask with three indentations spaced 120 degrees apart
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
The diameter at top is closely controlled to insure a fit with metal condensers.