BEAKERplus™ Beaker/Flask Combination, Kimble Chase
À propos de cet article
BEAKERplus is a combination Griffin beaker and Erlenmeyer flask and is ideal for mixing and for use with funnels or filters.
- Wide fluted rim with pour spout
- Narrow recessed neck minimizes splashing during vigorous agitation
- Easy-to-read white graduated scale is provided on all sizes from 150 to 1200 mL for measuring and/or mixing liquids
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
5 Options Available Below
Product Details & Documents
BEAKERplus is a combination Griffin beaker and Erlenmeyer flask and is ideal for mixing and for use with funnels or filters.
- Wide fluted rim with pour spout
- Narrow recessed neck minimizes splashing during vigorous agitation
- Easy-to-read white graduated scale is provided on all sizes from 150 to 1200 mL for measuring and/or mixing liquids
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Product Family Options
Informations sur le produit
- CapacitéIntervalle de graduationPlage de graduationHauteurØ ext.AvailabilityPrice
- 150 ml20 ml40 - 120 ml83 mm70 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 6Case of 6 - $408.85$408.85Prix promotionnel pour les nouveaux clientsCase of 6$408.85Supplier Number: 14035-150Spécifications:
More
Specifications
Cat. No.89218-794 - 250 ml50 ml50 - 200 ml107 mm70 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 6Case of 6 - $468.65$468.65Prix promotionnel pour les nouveaux clientsCase of 6$468.65Supplier Number: 14035-250Spécifications:
More
Specifications
Cat. No.89218-796 - 500 ml50 ml50 - 400 ml178 mm70 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 6Case of 6 - $518.70$518.70Prix promotionnel pour les nouveaux clientsCase of 6$518.70Supplier Number: 14035-500Spécifications:
More
Specifications
Cat. No.89218-798 - 1000 ml50 ml50 - 900 ml222 mm95 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 2Case of 2 - $195.50$195.50Prix promotionnel pour les nouveaux clientsCase of 2$195.50Supplier Number: 14035-1000Spécifications:
More
Specifications
Cat. No.89218-800 - 1200 ml50 ml50 - 1100 ml254 mm95 mmLog in pour la disponibilitéLog in pour la disponibilitéCase of 2Case of 2 - $224.77$224.77Prix promotionnel pour les nouveaux clientsCase of 2$224.77Supplier Number: 14035-1200Spécifications:
More
Specifications
Cat. No.89218-802