Wide Brooms, Soft/Stiff, Remco Products
Fournisseur: Remco Products
Certificats
À propos de cet article
This fully color-coded floor broom has two types of bristles to first help loosen stubborn dirt and move heavy debris, and then gather the finer particles
Block material is polypropylene; bristle material is polyester.
Certifications: FDA Compliant Raw Material.
Frequently Bought Together