ACTERO™ Yeast and Molds Enrichment Media, FoodChek Systems
Supplier: Foodchek
Certificates
About this item
Food and environmental samples require enrichment before pathogen testing, enabling any bacterial pathogen present to grow and be detected in test samples
Actero™ Universal Enrichment Media represent established formulations that are used in standard protocols employed in testing labs today.
Frequently Bought Together