Sterile Rabbit Sera, Rockland Immunochemicals
Catalogue # CAD109-00-0050
Fournisseur: Rockland Immunochemical
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
- Sterility:Sterile
- Description:Serum
- Size:50 mL
- Cat. no.:CAD109-00-0050
- Supplier No.:D109-00-0050
Spécifications
À propos de cet article
Sera is obtained from non-hemolyzed blood collected from healthy, fasted donors
Animals are maintained on an antibiotic-free diet. Sterilization is accomplished by Millipore filtration.
Ordering information: For heat-inactivated sera, contact your Avantor sales representative.