HyClone™ Equine Sera, HyClone products (Cytiva)
Supplier: Cytiva
Certificates
About this item
HyClone™ donor horse serum, designed to supplement cell growth, is collected from selected donor herds that receive regular veterinary inspection and care.
- Horses are bled in an enclosed facility that is specially equipped for aseptic collection
- Coggins tests for equine infectious anemia are routinely performed on each horse
- Serum filtered using sequential 100 nm pore-size rated filters
Supports hybridoma and antibody production. Meets specifications of \<=10EU/mL of endotoxin and \<=10mg/dL of hemoglobin. Filtered through triple 100nm pore-size rated filtration system. A biochemical profile is supplied.
Ordering information: All equine sera are packaged in PETG bottles.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA16777-026
- CA16777-030
- CA82013-590
- Storage temperature:
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C