ContactUsLinkComponent
Whole Horse Sera, Rockland Immunochemicals
Catalog # CAB307
Whole Horse Sera, Rockland Immunochemicals
Catalog # CAB307
Supplier Number:  B307
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Normal Whole Horse Serum
  • Size:
    10 mL
  • Cat. no.:
    CAB307
  • Supplier No.:
    B307
Specifications
About this item
The whole sera are prepared from normal sera by a multi-step process that includes delipidation and selective precipitation
All sera are derived from starting materials of North American origin and collection was performed in USDA inspected facilities. All materials have been inspected and certified to be free of disease and are suitable for exportation.