About this item
Defined fetal bovine serum (FBS) is the highest quality of FBS available and is very widely used by those cell culturist who have a concern for viral contaminants and require an extensive biochemical profile. Defined FBS is filtered through multiple 40 nm (0.04 µm) pore-size related filters, which are the most retentive filters used in commercial FBS production. Over 50 components are analysed and the results are included on the biochemical assay.
- Filtered through serial 40 nm pore-size rated filters
- Maximize cell culture growth
- U.S. sourced
Maximize cell culture growth with U.S.-sourced Cytiva HyClone Defined Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 16777-530
- CA16777-530
- CA16777-006
- 16777-006
- CA16777-002
- CA16750-030
- Storage temperature:
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C
- Less than –10 °C