Fluorescent Bulbs, Bulbtronics®
Supplier: Bulbtronics
Jump to:
About this item
Various fluorescent bulbs.
3 Options Available Below
Product Details & Documents
Various fluorescent bulbs.
Product Family Options
Product Information
- DésignationColorBulb LifeBulb TypeLongueurDiameterModèlePuissanceOEM NumberMaximum TemperatureAvailabilityPrice
- Eiko Fluorescent BulbBlacklight Blue7500 hLinear457.2 mm25.4 mmT815 WF15T8/BL—Log in for availabilityEach$19.26Supplier Number: 0017377
- Eiko Fluorescent Bulb, 295L, 60CRICool White7500 hLinear228.6 mm15.88 mmT56 W155054100KDiscontinued-Supplier Number: -
- Eiko Fluorescent Bulb, 825L, 60CRICool White7500 hLinear457.2 mm25.4 mmT815 W155214100KLog in for availabilityEach$10.71Supplier Number: 0017379
- General Electric Fluorescent Bulb, 135 L, 60CRICool White5000 hLinear152.4 mm15.88 mmT54 W100044100KDiscontinued-Supplier Number: -
- General Electric Fluorescent Bulb, 368 nm Spectral OutputBlacklight Blue5000 hLinear304.8 mm15.88 mmT58 W10077—Discontinued-Supplier Number: -
- General Electric Fluorescent Bulb, 365 nm Spectral OutputBlacklight Blue7500 hLinear457.2 mm25.4 mmT815 W35885—Discontinued-Supplier Number: -
- General Electric Fluorescent Bulb, 700 L, 75CRIDaylight7500 hLinear457.2 mm25.4 mmT815 W101346500KDiscontinued-Supplier Number: -
- General Electric Fluorescent Bulb, 825L, 60CRICool White7500 hLinear457.2 mm25.4 mmT815 W101424100KDiscontinued-Supplier Number: -
- General Electric Fluorescent Bulb, 1100L, 60CRI, with Energy SaverCool White12000 hLinear209.55 mm28.58 mmT922 W337744100KEach$228.73Case of 12$2,744.76Supplier Number: 0009028
- Hosobuchi Fluorescent BulbWhite2000 hRing-Light94.49 mm12.7 mmT46 W74B WW3000KDiscontinued-Supplier Number: -
- Osram Sylvania Fluorescent Bulb, 2180L, 60CRICool White7500 hLinear914.4 mm25.4 mmT830 WF30T8/CW4200KDiscontinued-Supplier Number: -
- Osram Sylvania Fluorescent Bulb, 6250L, 60CRICool White12000 hLinear1778 mm38.1 mmT1285 WF72T12/CW/HO4200KDiscontinued-Supplier Number: 0013137
- Ushio Fluorescent BulbDaylight2000 hRing-Light95.25 mm12.7 mmT46 W30001266250KDiscontinued-Supplier Number: -
- General Electric Fluorescent Bulb, 312 nm Spectral OutputDaylight—Linear304.8 mm15.88 mmT58 W26240—Discontinued-Supplier Number: -