ContactUsLinkComponent
Hei-VAP Industrial Safety Rotary Evaporators, 220 V
undefined
Hei-VAP Industrial Safety Rotary Evaporators, 220 V
  • Catalog No:
  • CA10018-640
  • CA10018-684
  • CA10018-648
  • CA10018-654
  • CA10018-636
  • CA10018-638
  • CA10018-682
  • Rotation speed:
  • 6 - 160 rpm
  • 6 - 160 rpm
  • 6 - 160 rpm
  • 6 - 160 rpm
  • 6 - 160 rpm
  • 6 - 160 rpm
  • 6 - 160 rpm
  • Bath capacity:
  • 24 L (water)/22 L (oil)
  • 24 L (water)/22 L (oil)
  • 24 L (water)/22 L (oil)
  • 24 L (water)/22 L (oil)
  • 24 L (water)/22 L (oil)
  • 24 L (water)/22 L (oil)
  • 24 L (water)/22 L (oil)
  • Dimensions:
  • 970×1600×690 mm
  • 970×1600×690 mm
  • 970×1600×690 mm
  • 970×1600×690 mm
  • 970×1600×690 mm
  • 970×1600×690 mm
  • 970×1600×690 mm
  • Temperature range:
  • 20...100 °C (water)/20...180 °C (oil)
  • 20...100 °C (water)/20...180 °C (oil)
  • 20...100 °C (water)/20...180 °C (oil)
  • 20...100 °C (water)/20...180 °C (oil)
  • 20...100 °C (water)/20...180 °C (oil)
  • 20...100 °C (water)/20...180 °C (oil)
  • 20...100 °C (water)/20...180 °C (oil)
  • Heating power:
  • 4000 W
  • 4000 W
  • 4000 W
  • 4000 W
  • 4000 W
  • 4000 W
  • 4000 W
  • Bath Ø:
  • 400 mm
  • 400 mm
  • 400 mm
  • 400 mm
  • 400 mm
  • 400 mm
  • 400 mm
Frequently Bought Together