VWR® Solvent Waste System
VWR® Solvent Waste System
VWR® Solvent Waste System
VWR® Solvent Waste System
This solvent waste system is designed to be an affordable solution for HPLC liquid waste disposal.
  • Activated carbon filter traps volatile organic compounds
  • Prevents chemicals from being inhaled
  • Closed-loop design
Each system is manufactured to safely collect waste from two HPLC machines.
This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The solvent waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety.
 : Each system includes a 4L, 5L, 10L, or 20L HDPE carboy, two 83B Versatile Caps (one open top for waste collection and one closed top for safe transport), six ports for 3.2mm (1/8”) O.D. waste tubes, one port for a carbon exhaust filter, one carbon exhaust filter and plugs for unused ports. Replacement fittings and carbon exhaust filters are also available.
  • Catalog No:
  • 10124-044
  • 10124-342
  • 10124-040
  • 10124-042
  • Environmentally Preferable:
  • Safer Human and Environmental Health|Waste Reducing
  • Safer Human and Environmental Health|Waste Reducing
  • Safer Human and Environmental Health|Waste Reducing
  • Safer Human and Environmental Health|Waste Reducing
Product Family Options
  • Désignation
    Matière
    Capacité
    Taille du bouchon
  • VWR® Solvent Waste Bottle Kit, 4L Bundle
    HDPE
    4 L (1.05 gal.)
    80 mm
    Each
    Each - $1,072.23
    $1,072.23
    Each
       $1,072.23
      Safer Human and Environmental Health|Waste Reducing
    Specifications
    Cat. No.
    10124-044
    Environmentally Preferable
    Safer Human and Environmental Health|Waste Reducing
      Safer Human and Environmental Health|Waste Reducing
  • VWR® Solvent Waste Carboy Kit, 5L Bundle
    HDPE
    5 L (1.32 gal.)
    80 mm
    Each
    Each - $1,182.16
    $1,182.16
    Each
       $1,182.16
      Safer Human and Environmental Health|Waste Reducing
    Specifications
    Cat. No.
    10124-342
    Environmentally Preferable
    Safer Human and Environmental Health|Waste Reducing
      Safer Human and Environmental Health|Waste Reducing
  • VWR® Solvent Waste Carboy Kit, 10L Bundle
    HDPE
    10 L (2.64 gal.)
    80 mm
    Each
    Each - $1,268.32
    $1,268.32
    Each
       $1,268.32
      Safer Human and Environmental Health|Waste Reducing
    Specifications
    Cat. No.
    10124-040
    Environmentally Preferable
    Safer Human and Environmental Health|Waste Reducing
      Safer Human and Environmental Health|Waste Reducing
  • VWR® Solvent Waste Carboy Kit, 20L Bundle
    HDPE
    20 L (5.28 gal.)
    80 mm
    Each
    Each - $1,275.25
    $1,275.25
    Each
       $1,275.25
      Safer Human and Environmental Health|Waste Reducing
    Specifications
    Cat. No.
    10124-042
    Environmentally Preferable
    Safer Human and Environmental Health|Waste Reducing
      Safer Human and Environmental Health|Waste Reducing
Accessories
  •  
    Description
  • 10124-338
    Replacement Carbon Filter
    Pack of 2 - $500.21
  •  
    Description
  • 10124-340
    Replacement Fittings (1/8" and 1/4")
    Each - $313.50
  •  
    Description
  • 10827-856
    EZwaste® System, 83 mm Cap, 6x 1/8" OD Tubing, and Filter
    Each - $912.54
  •  
    Description
  • 10827-858
    EZwaste® System, 83 mm Cap, 4x 1/8", 3x 1/4" OD Tubing and Filter
    Each - $882.99
  •  
    Description
  • 10827-860
    EZwaste® System, 83 mm Cap, 6x 1/8" OD Tubing, 1x 1/4 or 3/8" ID Hose Barb and Filter
    Each - $737.42
  •  
    Description
  • 10827-862
    EZwaste® System, 83 mm Cap, 4x 1/8", 3x 1/4" OD Tubing, 1x 1/4 or 3/8" ID Hose Barb and Filter
    Each - $865.25
  •  
    Description
  • 10827-864
    EZwaste® System, 83 mm Cap, 4x 1/8", 4x 1/4" OD Tubing and Filter
    Each - $892.18
  •  
    Description
  • 10827-866
    Replacement Exhaust Filter, Pack of 1
    Each - $390.19
  •  
    Description
  • 10827-868
    Replacement Exhaust Filter, Pack of 5
    Pack of 5 - $1,157.84