CritiCore ESD Stripe Cleanroom Hoods with Open Face Under Chin
Fournisseur: VWR International
Certificats
À propos de cet article
CritiCore’s reusable, controlled environment ESD stripe hoods are suitable for a variety of cleanroom applications, including pharmaceutical, biotechnology, and medical device manufacturing.
- Open face design under the chin
- Made with a high-density polyester calendered fabric that resists fluids and bacterial penetration
- Hoods feature a tunneled elastic back plus horizontal and vertical adjustment snaps
- Split-drape design with a front panel that is longer than the back
- All seams are laser-cut or pre-serged
- Thread is silicon-free
- Available in white and navy
- Sizes range from XS to 4XL
Designed for cleanroom applications up to and including Class 10 (ISO 4), these hoods also include a one percent carbon stripe for electrostatic dissipation.
Ordering information: Custom order product with extended lead time. Contact your Avantor sales representative for more information.
Spécifications
- Catalog No:
- 10120-398
- 10120-400
- 10120-378
- 10120-380
- 10120-402
- 10120-382
- 10120-404
- 10120-384
- 10120-406
- 10120-408
- 10120-386
- 10120-388
- 10120-390
- 10120-392
- 10120-394
- 10120-396
- Garment application:
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Cleanroom, Laboratory
- Risk area:
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- ESD
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Back fastening
- Fabric:
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- High-Density ESD (99% polyester, 1% carbon)
- Weight:
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.
- 2.53 oz./sq. yd.