Universal Plug Stoppers, Globe Scientific
Universal Plug Stoppers, Globe Scientific
Supplier:   Globe Scientific
Universal Plug Stoppers, Globe Scientific
Supplier:  Globe Scientific
Certificates
Universal Plug Stoppers, Globe Scientific
Universal Plug Stoppers, Globe Scientific
7 Options Available Below
Product Details & Documents
These universal plug stoppers fit most 10 mm, 12 mm, 13 mm, and 16 mm tubes
  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Add the convenience of 1 cap to fit multiple size tubes
These caps are available in 7 different colors.
Ordering information: Stoppers are packaged 1000 per bag.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Light Blue
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $91.62
    $91.62
    New Customer Promotional Price
    Pack of 1,000
       $91.62
    Supplier Number: 5529
  • Dark Blue
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $94.13
    $94.13
    New Customer Promotional Price
    Pack of 1,000
       $94.13
    Supplier Number: 5529DB
  • Green
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $94.13
    $94.13
    New Customer Promotional Price
    Pack of 1,000
       $94.13
    Supplier Number: 5529G
  • Lavender
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $94.13
    $94.13
    New Customer Promotional Price
    Pack of 1,000
       $94.13
    Supplier Number: 5529L
  • Red
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $93.68
    $93.68
    New Customer Promotional Price
    Pack of 1,000
       $93.68
    Supplier Number: 5529R
  • White
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $94.13
    $94.13
    New Customer Promotional Price
    Pack of 1,000
       $94.13
    Supplier Number: 5529W
  • Yellow
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $92.81
    $92.81
    New Customer Promotional Price
    Pack of 1,000
       $92.81
    Supplier Number: 5529Y