Order Entry
Canada
ContactUsLinkComponent
Mouse IgG1 Monoclonal Isotype Control [clone: MOPC-21] (PE (Phycoerythrin))
Catalogue # 10141-032
Fournisseur:  Tonbo Biosciences
undefined
Mouse IgG1 Monoclonal Isotype Control [clone: MOPC-21] (PE (Phycoerythrin))
Catalogue # 10141-032
Fournisseur:  Tonbo Biosciences
Supplier Number:  50-4714-U100

Spécifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Size:
    100 µg
  • Neutralisation:
    Yes
  • Storage temperature:
    Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
  • Tested applications:
    Flow Cytometry
  • Cat. no.:
    10141-032
  • Supplier No.:
    50-4714-U100

Spécifications

À propos de cet article

The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.

Type: Control
Antigen: MOPC21
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: