Mouse IgG1 Monoclonal Isotype Control [clone: MOPC-21] (FITC (Fluorescein Isothiocyanate))
Catalog # 10050-984
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:MOPC-21
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Size:100 µg
- Neutralisation:Yes
- Storage temperature:Store Undiluted at 2 to 8°C; Protect from Light; Do Not Freeze.
- Tested applications:Flow Cytometry
- Cat. no.:10050-984
- Supplier No.:35-4714-U100
Specifications
About this item
The MOPC-21 immunoglobulin is useful as an isotype-matched control. The MOPC-21 immunolglobulin has an unknown binding specificity and is used as an isotype control for mouse IgG1 antibodies.
Type: Control
Antigen: MOPC21
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: