Duracell® Lithium Batteries, Bulbtronics
Hazmat
Non-rechargeable.
Frequently Bought Together
VWR® Self-Seal Sterilization Pouches
: VWR International
Gauze Pads, DUKAL™ Corporation
: Dukal
Alconox® Powdered Precision Cleaners
: Alconox
VWR® Blood Bank Saline Solutions
: VWR International
Leakbuster™ Specimen Containers
: Starplex
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
: MilliporeSigma
Histoplex™ Histology Containers, Starplex®
: Starplex
Glutaric acid ≥99.0% (by GC, titration analysis)
: TCI America
EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) 99%
: Thermo Scientific Chemicals
1,4-Diacetylbenzene ≥99.0%
: TCI America