Isopropanol, Contec® | Avantor
Isopropanol, Contec®
Isopropanol, Contec®
Supplier:  CONTEC, INC. MX
Certificates
Isopropanol, Contec®
Isopropanol, Contec®
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Contec® Isopropanol contains USP grade isopropanol (IPA) and USP grade purified water. Filtered to 0.2 μm, bottles are double-bagged to assure product quality. Each bottle has a lot number code and expiration date for easy record keeping.
  • Contains USP grade isopropanol which evaporates and leaves extremely low residues
  • Filtered through 0.2 μm in-line filter to eliminate the need for in-house filtration
  • Meets USP grade purified water
A convenient trigger sprayer with settings for stream or mist spraying is included with the 32 oz. bottles. The stream setting is ideal for targeting specific hard to reach areas, while the mist setting is used to cover larger surfaces. Each leakproof bottle is induction sealed, capped, and shipped to eliminate the occurrence of leakage. Product applications include surface cleaning, wiping gloved hands in sterile suites or during gowning, and wiping down products and equipment for pass-through to controlled environments.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Packaging
    Availability
    Price
  • Contec® 65% IPA, Nonsterile 65% USP Grade Isopropanol
    3.8 L (1 gal.)
    4 Bottles/Case
    Discontinued
    Supplier Number: -
  • Contec® 70% IPA, Nonsterile 70% USP Grade Isopropanol
    0.9 L (32 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $426.00
    $426.00
    Case of 12
       $426.00
    Supplier Number: SB327030
  • Contec® 70% IPA, Nonsterile 70% USP Grade Isopropanol
    3.8 L (1 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4 (4l)
    Case of 4 (4l) - $285.52
    $285.52
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Each(4l)
    Each(4l) - $285.52
    $285.52
    Prix promotionnel pour les nouveaux clients
    Case of 4 (4l)
       $285.52
    Each(4l)
       $285.52
    Supplier Number: SB1287030
  • Contec® 70% IPA, Nonsterile 70% USP Grade Isopropanol, trigger spray installed
    0.47 L (16 oz.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12 (1/Itm/Bx)
    Case of 12 (1/Itm/Bx) - $152.22
    $152.22
    Prix promotionnel pour les nouveaux clients
    Case of 12 (1/Itm/Bx)
       $152.22
    Supplier Number: SBT167030
  • Contec® 99% IPA, Non-sterile USP grade Isopropanol
    3.8 l (1 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $215.00
    $215.00
    Case of 12
       $215.00
    Supplier Number: SB128100