Science Take-Out® Enzymes And Lactose Intolerance
Supplier: SCIENCE TAKE-OUT, LLC
Certificates
About this item
How Do Enzymes Help People With Lactose Intolerance?
- Available As A Single Kit With Materials For 1 Student Or Group
- Unassembled 10 Pack For Additional Savings
- No Additional Equipment Needed
How does the dietary supplement LACTAID work to help people with lactose intolerance? Model the action of the enzyme lactase. Conduct tests to determine whether LACTAID, an enzyme supplement, digests lactose. Design and conduct an experiment to determine if acid interferes with the action of the enzymes in LACTAID. This complete experiment setup contains all materials and does not require lab equipment.
Frequently Bought Together