Vernier® Ethanol Sensor
Vernier ethanol sensor (BTA)
Catalog # CA470112-922
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: ETH-BTA
Vernier® Ethanol Sensor
Catalog # CA470112-922
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
Measure the concentration of ethanol in air above an aqueous sample.
- Wired connection with BTA plug
This sensor can be used in a wide range of tests and experiments. It can be used to determine the rate of ethanol production during fermentation and can also be used to measure the discrete amount of ethanol in a given sample.
Ordering information: LabQuest® (2, 3, Mini) interface is required but not included.
Spécifications
- Désignation:Vernier ethanol sensor (BTA)
- Cat. No.:CA470112-922
- Response time:60 s
- Range:0 - 3%