Vernier® Electrode Support
Vernier electrode support
Catalog # CA470112-920
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: ESUP
Vernier® Electrode Support
Catalog # CA470112-920
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
New Product
Jump to:
Product Details & Documents
Our Electrode Support is a great complement to the Vernier Stir Station, as well as a perfect holder for many sensors. It is built to connect to all standard ring-stand posts; its large-handled locking nut keeps your sensors firmly in place.
- The larger opening (13 mm Ø) is perfect for our pH, ISE, Conductivity and ORP sensors
- The smaller opening (5 mm) is designed to hold our Stainless Steel Temperature Probe or the Vernier Go!Temp
Spécifications
- Désignation:Vernier electrode support