Vernier® Energy Sensor | Avantor
Vernier® Energy Sensor
Vernier energy sensor (BTA), Source input potential: ±30 V; Source input current: ±1000 mA
Catalog # CA470116-632
New Product
Fournisseur #:  VES-BTA
Vernier® Energy Sensor
Catalog # CA470116-632
New Product
Vernier® Energy Sensor
Vernier® Energy Sensor
Vernier energy sensor (BTA), Source input potential: ±30 V; Source input current: ±1000 mA
Price
$206.09
Log in pour la disponibilité
Each
La quantité minimale de commande est de 1
Product Details & Documents
Easily quantify voltage, current, power, and energy output of small wind turbines and solar panels.
  • Wired connection with BTA plug
Setting up a circuit to study renewable energy systems such as wind turbines or solar panels is easy with the Vernier Energy Sensor. Simply connect a source such as a small wind turbine or solar panel to one side and a load such as the Variable Load or a resistor on Vernier Resistor Board to the other side.
The Energy Sensor measures the current and voltage of the system. Data-collection and analysis software, such as Logger Pro or LabQuest® app can then calculate the power and energy output.
Ordering information: LabQuest® (2, 3, Mini) interface is required but not included.
Spécifications
  • Désignation:
    Vernier energy sensor (BTA)
  • Range:
    Source input potential: ±30 V; Source input current: ±1000 mA