96-Well Microplates, Polystyrene, With Lids, Greiner Bio-One
Catalog # 89089-578
Supplier: Greiner Bio-One
Product available on GSA Advantage®
Specifications
- Color:Clear
- Material:Polystyrene
- Cat. no.:89089-578
- Bottom Style:Flat bottom
- Supplier No.:655184
Specifications
About this item
96 well plate manufactured from clear polystyrene.
- Clear polystyrene construction
- Available in a variety of molding options
- Chimney style option helps prevent cross-contamination
Plates are available with chimney style, flat bottom, and sterile with lid.
Ordering information: Plates are packaged 8 per pack, 4 packs per case (32 plates per case).