Vernier® Anemometer
Vernier anemometer (BTA)
Catalog # CA470112-970
New Product
Supplier: VERNIER CANADA SCIENCE EDUCATION IN
Fournisseur #: ANM-BTA
Jump to:
Product Details & Documents
Measure wind speed in dynamic environments.
- Fits in your palm
- Standard camera mount
The Vernier Anemometer is an impeller-type anemometer that measures wind speed in the range of 0.5 to 30 m/s (1 to 67 mph). The Anemometer fits in your palm for wind study measurements in the field. A standard camera mount on the back and an accessory rod allows you to position it in wind tunnels or in front of fans for wind turbine experiments.
For educational use only.
Spécifications
- Désignation:Vernier anemometer (BTA)
- Cat. No.:CA470112-970
- Range:0.5 - 30 m/s
- Resolution:1.2×10 - 2 m/s