TechNiCleaned Class 10 Nitrile Cleannrom Gloves, Techniglove
TechNiCleaned Class 10 Nitrile Cleannrom Gloves, Techniglove
TechNiCleaned Class 10 Nitrile Cleannrom Gloves, Techniglove
Supplier:  Techniglove
Certificates
TechNiCleaned Class 10 Nitrile Cleannrom Gloves, Techniglove
TechNiCleaned Class 10 Nitrile Cleannrom Gloves, Techniglove
18 Options Available Below
Product Details & Documents
TechniCleaned Ambidextrous 12" Class 10 powder free nitrile gloves are designed for the most critical applications and are manufactured in 3 colors: White, Blue and Green.
  • Class 10
  • 100% Nitrile
  • 12" ambidextrous
  • Powder-free
  • For use in semiconductor, disc drive, flat panel, electronic, and pharmaceutical manufacturing
Techniglove products contain no natural rubber or silicone, and meet the most stringent requirements for particles and extractables. The proprietary Microtexture Formers offer an exceptional fit and a soft, supple feel. Exclusive On-line Batch Testing is available for this product.
Multiple colors allow coding by size, coding by product, coded double gloving and many other opportunities. Suitable for use in semiconductor, disc drive, flat panel, electronic and pharmaceutical manufacturing.
Certifications: COC available on request.
Ordering information: Packaged 100 gloves per poly-sealed cleanroom bag, 10 bags per case.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89414-958
  • 89414-960
  • 89414-962
  • 89414-964
  • 89414-966
  • 89414-968
  • 89414-970
  • 89414-972
  • 89414-974
  • 89414-976
  • 89414-978
  • 89414-980
  • 89414-982
  • 89414-984
  • 89414-986
  • 89414-988
  • 89414-990
  • 89414-992
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Finish:
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
  • Microtextured Fingertips
Spécifications
Product Family Options
Informations sur le produit
  • Taille du gant
    Glove Color
    Longueur
    Épaisseur
    Availability
    Price
  • XS
    White
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1200W
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-958
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • S
    White
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $926.83
    $926.83
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $926.83
    Supplier Number: TN1201W
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-960
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • M
    White
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $926.83
    $926.83
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $926.83
    Supplier Number: TN1202W
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-962
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • L
    White
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,007.12
    $1,007.12
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,007.12
    Supplier Number: TN1203W
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-964
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XL
    White
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,235.32
    $1,235.32
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,235.32
    Supplier Number: TN1204W
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-966
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XXL
    White
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.87
    $1,136.87
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.87
    Minimum order quantity is 1
    Supplier Number: TN1205W
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-968
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XS
    Blue
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1200B
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-970
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • S
    Blue
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,235.32
    $1,235.32
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,235.32
    Supplier Number: TN1201B
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-972
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • M
    Blue
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,235.32
    $1,235.32
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,235.32
    Supplier Number: TN1202B
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-974
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • L
    Blue
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1203B
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-976
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XL
    Blue
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,235.32
    $1,235.32
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,235.32
    Supplier Number: TN1204B
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-978
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XXL
    Blue
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1205B
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-980
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XS
    Green
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1200G
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-982
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • S
    Green
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1201G
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-984
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • M
    Green
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,027.10
    $1,027.10
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,027.10
    Supplier Number: TN1202G
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-986
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • L
    Green
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,027.10
    $1,027.10
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,027.10
    Supplier Number: TN1203G
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-988
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XL
    Green
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,235.32
    $1,235.32
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,235.32
    Supplier Number: TN1204G
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-990
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips
  • XXL
    Green
    30.5 cm (12")
    5 mil.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,136.84
    $1,136.84
    Prix promotionnel pour les nouveaux clients
    Case of 1,000
       $1,136.84
    Supplier Number: TN1205G
    Spécifications:
    More
    Specifications
    Cat. No.
    89414-992
    Ambidextrous/Hand-specific
    Ambidextrous
    Finish
    Microtextured Fingertips