Corning® Bar Coded Storage Tubes, Corning
Corning® Bar Coded Storage Tubes, Corning
 :   Corning
Corning® Bar Coded Storage Tubes, Corning
 :  Corning
Corning® Bar Coded Storage Tubes, Corning
Corning® Bar Coded Storage Tubes, Corning
PP. The 1D/2D storage line is an innovative product with both a linear 1D and 2D barcodes etched on each tube.
  • Maximum information, 14×14 dot 2D barcode
  • Barcode stability, laser-etched, not an attached label
  • Compound compatibility, polypropylene construction throughout, making it inert
Product Family Options
96 Tubes
  • Désignation
    Volume
    Packaging
  • 96 1D/2D barcoded storage tubes, without screw caps, Natural
    1.3 ml
    bulk packed
    Case of 960
    Case of 960 - $1,580.00
    $1,580.00
    Case of 960
       $1,580.00
     : 8500
  • 96× 1D/2D barcoded storage tubes, without screw caps, Natural
    1.3 ml
    racked with cover
    Case of 960
    Case of 960 - $1,740.00
    $1,740.00
    Case of 960
       $1,740.00
     : 8501
  • 96× 1D/2D barcoded storage tubes, with screw caps, Orange
    1.3 ml
    bulk packed
    Case of 960
    Case of 960 - $2,280.00
    $2,280.00
    Case of 960
       $2,280.00
     : 8502
  • 96× 1D/2D barcoded storage tubes, with screw caps, Orange
    1.3 ml
    racked with cover
    Case of 960
    Case of 960 - $2,570.00
    $2,570.00
    Case of 960
       $2,570.00
     : 8503
384 Tubes
  • Désignation
    Volume
    Packaging
  • 384× 2D barcoded storage tubes, round, without plug caps, Black
    110 µl
    racked
    Case of 7,680
    Case of 7,680 - $4,060.00
    $4,060.00
    Case of 7,680
       $4,060.00
     : 8509
  • 384× 2D barcoded storage tubes, square, without plug caps, Black
    120 µl
    racked
    Case of 7,680
    Case of 7,680 - $4,060.00
    $4,060.00
    Case of 7,680
       $4,060.00
     : 8510
Accessories
  •  
    Description
  • 89168-514
    96 screw caps for storage tubes 1,3 ml, with O-Ring, Orange, for no. 8500
    Case of 4,800 - $5,060.00
  •  
    Description
  • 89168-516
    96 storage tube screw cap, with O-Ring, on mat, clear with cover, Orange, for no. 8500
    Case of 4,800 - $5,880.00
  •  
    Description
  • 89168-518
    Thermoplastic elastomer (TPE) 96 storage tube septum cap, White, for no. 8500
    Case of 9,600 - $901.00
  •  
    Description
  • 89168-520
    Thermoplastic elastomer (TPE) 96 storage tube septum cap on mat, White, for no. 8500
    Case of 4,800 - $1,480.00
  •  
    Description
  • 89168-522
    Empty racks with lids for 96 screw capped storage tubes no. 8502, Natural
    Case of 10 - $145.00
  •  
    Description
  • 89168-530
    384 storage tube plug cap on mats for round tube no. 8509, 384 caps per mat, Orange
    Case of 38,400 - $7,100.00
  •  
    Description
  • 89168-528
    384 Storage Tube Plug Cap,Orange, for no. 8509
    Case of 38,400 - $5,080.00
  •  
    Description
  • 89168-532
    Empty racks with lids for 384 round or square storage tubes, Blue, for nos. 8509 and 8510
    Case of 20 - $366.00