VortaMix Mini Vortexer, Argos Technologies
VortaMix Mini Vortexer, Argos Technologies
Supplier:   Antyila Scientific
VortaMix Mini Vortexer, Argos Technologies
Supplier:  Antyila Scientific
Certificates
VortaMix Mini Vortexer, Argos Technologies
2 Options Available Below
Product Details & Documents
The Vortamix Mini Vortexer from Argos Technologies is an unbelievably compact, yet powerful instrument engineered to mix tubes quickly and easily
The standard unit comes with a red rubber gasket cap, but blue, green and purple colored caps are offered separately
Despite its small size and weight, the Vortamix offers an excellent mixing speed of 3,000 rpm.
The vortexing motion is activated by placing a tube on the gasket cap and then pressing down lightly. The innovative design of the gasket cap allows centrifuge and test tubes up to 30 mm in diameter to be mixed with very little effort – including 50mL conical tubes.
Certifications: The Vortamix is CE marked and has and IP rating of DIN EN 60529.
Ordering information: This new instrument comes with a 2 year warranty.
Specifications for Product Family
Specifications
  • Catalog No:
  • 10118-092
  • 10118-094
  • Speed:
  • 3,000 rpm
  • 3,000 rpm
  • Weight:
  • 0.55 kg
  • 0.55 kg
  • Operating temperature:
  • 5...40 °C
  • 5...40 °C
  • Dimensions:
  • 95 x 85 mm
  • 96 x 85 mm
Specifications
Product Family Options
Product Information
  • Désignation
    Orbit
    Availability
    Price
  • Argos Mini Vortexer, US Adapter
    4.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $587.57
    $587.57
    New Customer Promotional Price
    Each
       $587.57
    Supplier Number: 04397-27
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    10118-092
    Speed
    3,000 rpm
    Weight
    0.55 kg
    Operating temperature
    5...40 °C
    Dimensions
    95 x 85 mm
    Small Business Enterprise
  • Argos Mini Vortexer, International Adapter
    4.5 mm
    Log in for availability
    Log in for availability
    Each
    Each - $568.92
    $568.92
    New Customer Promotional Price
    Each
       $568.92
    Supplier Number: 04397-28
    Specifications:
    Small Business Enterprise
    More
    Specifications
    Cat. No.
    10118-094
    Speed
    3,000 rpm
    Weight
    0.55 kg
    Operating temperature
    5...40 °C
    Dimensions
    96 x 85 mm
    Small Business Enterprise
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Argos Head Adapter, Red
    Argos Head Adapter, Red
    10118-096
    Log in for availability
    Each - $29.32
Product Information
  • Catalog #
    Availability
    Price
  • Argos Head Adapter, Blue
    Argos Head Adapter, Blue
    10118-098
    Log in for availability
    Each - $30.64
Product Information
  • Catalog #
    Availability
    Price
  • Argos Head Adapter, Purple
    Argos Head Adapter, Purple
    10118-102
    Log in for availability
    Each - $30.64