Corning® Solid Black and White Polystyrene Microplates, 384 Well
Corning® Solid Black and White Polystyrene Microplates, 384 Well
 :   Corning
Corning® Solid Black and White Polystyrene Microplates, 384 Well
 :  Corning
Corning® Solid Black and White Polystyrene Microplates, 384 Well
Corning® Solid Black and White Polystyrene Microplates, 384 Well
PS. Designed to reduce well-to-well crosstalk during fluorescent and luminescent assays.
  • Square well top for easier robotic pipetting
  • Round bottom for better Z factor and minimized trapped air
  • Conical well molded in the shape of a light cone for efficiency
  • Black walls reduce well-to-well crosstalk and autofluorescence during fluorescent assays
High binding surface is a general purpose surface that binds medium (>10kD) and large biomolecules possessing ionic groups and/or hydrophobic regions. Binding capacity is approximately 400 to 500ng IgG/cm.
Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions. Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface (binding capacity of approximately 100 to 200ng IgG/cm²).
Nonbinding surface (NBS™) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions. Ideal for reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise.
  • Catalog No:
  • 89511-310
  • CA89089-786
  • 89511-312
  • CA89089-798
  • 89511-314
  • 89511-316
  • 89511-318
  • 89089-856
  • 89089-858
  • 89089-860
  • 89089-868
  • 89185-546
  • 89089-870
  • 89203-194
  • 89203-192
  • 89089-788
  • 89089-794
  • 89089-796
  • 89089-800
  • 89176-442
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
  • RECs, ISO 14001 - Kennebunk
Product Family Options
  • Désignation
    Total Well Volume
    Well Volume
    Working Well Volume
    Color
    Sterility
    Surface Treatment
    Well Shape
    Packaging
  • 384 Well Low Volume Microplate, Without Lid
    35 µl
    50 µl
    5 - 40 µl
    Black
    Non sterile
    High binding
    Round-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,320.00
    $1,320.00
    Case of 50
       $1,320.00
     : 4510
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89511-310
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates with bar code
    112 µl
    112 µl
    20 - 80 µl
    Black
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $949.00
    $949.00
    Case of 50
       $949.00
     : 3575BC
    Specifications
    Cat. No.
    CA89089-786
    Environmentally Preferable
    Environmental certification
  • 384 Well Low Volume Microplate, Without Lid
    35 µl
    35 µl
    2 - 25 µl
    Black
    Non sterile
    Untreated
    Round-Bottom
    10/Bag
    Case of 50
    Case of 50 - $674.90
    $674.90
    Case of 50
       $674.90
     : 4511
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89511-312
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates with bar code
    112 µl
    112 µl
    20 - 80 µl
    White
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,010.00
    $1,010.00
    Case of 50
       $1,010.00
     : 3574BC
    Specifications
    Cat. No.
    CA89089-798
    Environmentally Preferable
    Environmental certification
  • 384 Well Low Volume Microplate, Without Lid
    35 µl
    50 µl
    5 - 40 µl
    White
    Non sterile
    Untreated
    Round-Bottom
    10/Bag
    Case of 50
    Case of 50 - $750.55
    $750.55
    Case of 50
       $750.55
     : 4512
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89511-314
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 384 Well Low Volume Microplate, Without Lid
    35 µl
    50 µl
    5 - 40 µl
    White
    Non sterile
    NBS™
    Round-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,003.00
    $1,003.00
    Case of 50
       $1,003.00
     : 4513
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89511-316
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 384 Well Low Volume Microplate, Without Lid
    35 µl
    50 µl
    2 - 20 µl
    Black
    Non sterile
    NBS™
    Round-Bottom
    10/Bag
    Case of 50
    Case of 50 - $918.00
    $918.00
    Case of 50
       $918.00
     : 4514
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89511-318
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Corning® 384 Well Low Volume Black Flat Bottom Polystyrene NBS™ Microplate
    50 µl
    50 µl
    5 - 40 µl
    Black
    Non sterile
    NBS™
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $952.49
    $952.49
    Case of 50
       $952.49
     : 3820
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-856
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Corning® 384 Well Low Volume Black Flat Bottom Polystyrene Not Treated Microplate
    50 µl
    50 µl
    5 - 40 µl
    Black
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $909.50
    $909.50
    Case of 50
       $909.50
     : 3821
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-858
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Corning® Low Volume 384 Well Black Flat Bottom Polystyrene TC-Treated Microplate, With Lid
    50 µl
    50 µl
    5 - 40 µl
    Black
    Sterile
    Tissue culture (TC)
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,096.50
    $1,096.50
    Case of 50
       $1,096.50
     : 3822
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-860
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Corning® Low Volume 384 Well White Flat Bottom Polystyrene NBS™ Microplate
    50 µl
    50 µl
    5 - 40 µl
    White
    Non sterile
    NBS™
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $977.50
    $977.50
    Case of 50
       $977.50
     : 3824
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-868
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Corning® Low Volume 384 Well White Flat Bottom Polystyrene Non Treated Microplate
    112 µl
    112 µl
    5 - 40 µl
    White
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $935.00
    $935.00
    Case of 50
       $935.00
     : 3825
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89185-546
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Corning® Low Volume 384 Well White Flat Bottom Polystyrene TC-Treated Microplate, With Lid
    50 µl
    50 µl
    5 - 40 µl
    White
    Sterile
    Tissue culture (TC)
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,079.50
    $1,079.50
    Case of 50
       $1,079.50
     : 3826
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-870
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Low volume plate, with bar code
    50 µl
    50 µl
    5 - 40 µl
    White
    Non sterile
    NBS™
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,156.00
    $1,156.00
    Case of 50
       $1,156.00
     : 3824BC
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89203-194
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Low volume plates, with bar code
    50 µl
    50 µl
    5 - 40 µl
    Black
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $1,079.50
    $1,079.50
    Case of 50
       $1,079.50
     : 3821BC
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89203-192
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates
    112 µl
    112 µl
    20 - 80 µl
    Black
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $547.20
    $547.20
    Case of 50
       $547.20
     : 3573
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-788
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates
    112 µl
    112 µl
    20 - 80 µl
    White
    Non sterile
    High binding
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $563.55
    $563.55
    Case of 50
       $563.55
     : 3576
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-794
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates
    112 µl
    112 µl
    20 - 80 µl
    White
    Non sterile
    Untreated
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $839.80
    $839.80
    Case of 50
       $839.80
     : 3574
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-796
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates with lid
    112 µl
    112 µl
    20 - 80 µl
    White
    Sterile
    Tissue culture (TC)
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $600.10
    $600.10
    Case of 50
       $600.10
     : 3570
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89089-800
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Solid plates with lid and bar code
    112 µl
    112 µl
    20 - 80 µl
    White
    Sterile
    Tissue culture (TC)
    Flat-Bottom
    10/Bag
    Case of 50
    Case of 50 - $733.55
    $733.55
    Case of 50
       $733.55
     : 3570BC
    Product available on GSA Advantage®
      Low Manufacturing Impact
    Specifications
    Cat. No.
    89176-442
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Kennebunk
    Product available on GSA Advantage®
      Low Manufacturing Impact