Synthware Condensers for Rotary Evaporators, Upright Style, Kemtech America
Catalog # 60001-180
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Condenser for rotary evaporators, vertical style
- Environmentally Preferable:
- Cat. no.:60001-180
- Supplier No.:C900200
Specifications
About this item
For vertical style of Heifdorfe.
- Upright design
- Heavy wall borosilicate construction
- Designed for Heidorfe roto vap