Synthware Distribution Receiver, Vertical Orientation, Kemtech America
Catalogue # 60002-138
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint:24/40
- Description:Vacuum adapter
- Environmentally Preferable:
- Cat. no.:60002-138
- Supplier No.:A592440
Spécifications
À propos de cet article
Rotating distillation receiver allows four fractions to be collected.
- Vertical orientation
- Heavy wall borosilicate construction
- Lower drip tip
- Hose connection side arm
Vacuum adapter has a vertical orientation.