Synthware Vacuum Adapter, 90° with Outer Joint, Kemtech America
Fournisseur: Kemtech America
Certificats
À propos de cet article
Vacuum adapter features a 90° angle hose connection.
- 90° angle
- Top hose connection
- Outer [ST] joint
Featuring a 90° angle hose connection on the top and an outer joint.
Frequently Bought Together