Synthware Flasks for Solvent Transfer/Storage, Kemtech America
Flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
- High Vacuum Valve included
- Various sizes available
- 14/20 and 24/40 ground glass joint available
Heavy wall synthware straus flask is commonly used for storing dired and degassed solvents.
Frequently Bought Together