ContactUsLinkComponent
Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)
Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
   Sustainable Options Available
Certificates
undefined
Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)
Supplier:  Cytiva
   Sustainable Options Available
Specifications
  • Catalog No:
  • CA76361-280
  • CA76382-924
  • Environmentally Preferable:
  • Waste Reducing
  • Max. operating pressure:
  • 30 psi
  • 30 psi
  • Max. temperature:
  • 34 °C
  • 34 °C