Extracta™ DNA Prep for PCR, Quantabio | Avantor
Extracta™ DNA Prep for PCR, Quantabio
Extracta™ DNA Prep for PCR, Quantabio
Supplier:  Quantabio
Certificates
Extracta™ DNA Prep for PCR, Quantabio
3 Options Available Below
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Extracta DNA Prep for PCR is a two-component reagent kit for rapid extraction of PCR-ready genomic DNA from a variety of tissues. Samples are processed in less than 30 minutes with minimal hands-on time and technical skill.
  • Simple, fast protocol
  • Compatible with laboratory automation
  • DNA is suitable for qPCR, High Resolution Melt Analysis, endpoint PCR and other applications
  • Works with wide range of tissue including hair, buccal swabs and mouse tails
  • Optional 10 minute protocol for high-throughput applications
Quanta's Extracta sample preparation system is a system that can be scaled to process various amounts of solid tissue in a simple and fast single-tube procedure. DNA may be purified from many tissue types and is suitable for use in sensitive downstream applications such as PCR, qPCR and High Resolution Melt Analysis.
The Extracta™ DNA prep for PCR consists of a two component reagent kit for rapid and efficient extraction of PCR-ready genomic DNA from mammalian tissues. The kit has been demonstrated to work with mouse tail biopsies, ear punches and a variety of human tissue samples including hair, buccal cells and saliva. The genomic DNA is suitable for sensitive downstream PCR applications including end-point PCR, High Resolution Melt Analysis (HRM) and quantitative real time PCR (qPCR) without requiring any additional clean-up. In addition, the extracted DNA may be used in multiplexed PCR applications such as transgene or knock-out analyses. Tissue extractions can be performed in tubes, plates or deep well blocks to allow for adaptation to workflow and automation on liquid handling workstations.
Ordering information: DNA Prep kit includes extraction reagent and stabilization buffer.
Documents
Specifications for Product Family
Spécifications
Spécifications
  • Catalog No:
  • 76047-142
  • CA76047-142
  • CA97065-350
  • CA97065-352
  • Description:
  • Extracta™ DNA prep for PCR
  • Extracta™ DNA prep for PCR
  • Extracta™ DNA prep for PCR
  • Extracta™ DNA prep for PCR
  • Target molecule:
  • DNA
  • DNA
  • DNA
  • DNA
  • Refine target molecule:
  • Total DNA
  • Total DNA
  • Total DNA
  • Total DNA
  • Sample type:
  • Cultured cells|Human/animal cells and tissues
  • Cultured cells|Human/animal cells and tissues
  • Cultured cells|Human/animal cells and tissues
  • Cultured cells|Human/animal cells and tissues
  • Method / Format:
  • Extraction for direct PCR
  • Extraction for direct PCR
  • Extraction for direct PCR
  • Extraction for direct PCR
  • Prep size:
  • Scalable
  • Scalable
  • Scalable
  • Scalable
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Extracta™ DNA prep for PCR
    2.5 ml
    Discontinued
    Supplier Number: 95091-002
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    76047-142
    Description
    Extracta™ DNA prep for PCR
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable
  • Extracta™ DNA prep for PCR
    2.5 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.01
    $32.01
    Each
       $32.01
    La quantité minimale de commande est de 1
    Supplier Number: 95091-002
    L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76047-142
    Description
    Extracta™ DNA prep for PCR
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable
  • Extracta™ DNA prep for PCR
    25 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $284.77
    $284.77
    Each
       $284.77
    Supplier Number: 95091-025
    Spécifications:
    More
    Specifications
    Cat. No.
    CA97065-350
    Description
    Extracta™ DNA prep for PCR
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable
  • Extracta™ DNA prep for PCR
    250 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,370.85
    $2,370.85
    Each
       $2,370.85
    Supplier Number: 95091-250
    Spécifications:
    More
    Specifications
    Cat. No.
    CA97065-352
    Description
    Extracta™ DNA prep for PCR
    Target molecule
    DNA
    Refine target molecule
    Total DNA
    Sample type
    Cultured cells|Human/animal cells and tissues
    Method / Format
    Extraction for direct PCR
    Prep size
    Scalable