50/84 Place Fusion Furnace, Furnace and Assay Supplies
Fournisseur: KLEN INTERNATIONAL (74) PTY LTD.
Certificats
À propos de cet article
These 50/84-place fusion furnaces are available with LPG, natural gas, or diesel fuel source options
- Suitable for large labs; 400-1200 samples per day including standards
- Base construction uses high-quality 100 x 100 RHS
- Furnace body construction is durable 5mm laser-cut plate
- High quality refractory brick and insulation
- Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
- Pneumatic door controls with foot pedal controller—available with an optional hand valve controller
This furnace has a fan forced dual burner system, with an automatic ignition system. Temperature is monitored and modulated automatically by a Type K thermocouple system and Omron temperature controller, pre-set for a maximum temperature of 1200 degrees Celsius. This controller is simple to operate, and has a 2-level display (PV and SV), that shows actual temperature as well as the temperature the furnace is set to. The door has a pneumatic ram opening system with an easy to use foot pedal control.
Crucibles should be 50 x 50g or 84 x 40g.
Ordering information: For optional equipment, contact your sales representative.
Caution: Please note that a minimum of 1 meter clearance around the furnace is required.
Spécifications
- Catalog No:
- CA11023-250
- CA76047-132
- CA76047-134
- CA11023-246
- CA11023-248
- Shipping dimensions:
- 209 x 169 x 208 cm
- 209 x 169 x 208 cm
- 209 x 169 x 208 cm
- 209 x 169 x 208 cm
- 209 x 169 x 208 cm
- Shipping weight:
- 2200 kg
- 2200 kg
- 2200 kg
- 2200 kg
- 2200 kg