Alpha Refrigerated Circulators, Lauda-Brinkmann
À propos de cet article
The Lauda-Brinkmann Alpha heating and cooling circulators provide a temperature range from -25 to 100 °C, depending on the exact model. This temperature range was selected with a focus of addressing most of the common temperature control applications within the laboratory. This entry level equipment range is made possible by minimizing any unnecessary features, with the focus on reliability and user-friendliness.
The Alpha heating and cooling circulators are suitable for operation with non-flammable liquids (water, water/glycol) and for both internal and external temperature control tasks. A one point calibration of the thermostats can be carried out by the user. Stainless steel bath.
3 Options Available Below
Product Details & Documents
The Lauda-Brinkmann Alpha heating and cooling circulators provide a temperature range from -25 to 100 °C, depending on the exact model. This temperature range was selected with a focus of addressing most of the common temperature control applications within the laboratory. This entry level equipment range is made possible by minimizing any unnecessary features, with the focus on reliability and user-friendliness.
The Alpha heating and cooling circulators are suitable for operation with non-flammable liquids (water, water/glycol) and for both internal and external temperature control tasks. A one point calibration of the thermostats can be carried out by the user. Stainless steel bath.
Specifications for Product Family
Spécifications
- Catalog No:
- 97039-888
- 97039-890
- 97039-892
- Pump flow rate:
- 15 L/min. (Max.)
- 15 L/min. (Max.)
- 15 L/min. (Max.)
- Pump pressure:
- .2 Bar (Max.)
- .2 Bar (Max.)
- .2 Bar (Max.)
Spécifications
Product Family Options
Informations sur le produit
- ModèlePlage de températureElectricalPuissance cryoHeating PowerAvailabilityPrice
- Supplier Number: L000650Spécifications:
More
Specifications
Cat. No.97039-888Pump flow rate15 L/min. (Max.)Pump pressure.2 Bar (Max.) - Supplier Number: L000651Spécifications:
More
Specifications
Cat. No.97039-890Pump flow rate15 L/min. (Max.)Pump pressure.2 Bar (Max.) - Supplier Number: L000652Spécifications:
More
Specifications
Cat. No.97039-892Pump flow rate15 L/min. (Max.)Pump pressure.2 Bar (Max.)