Alpha Heating Circulators, Lauda-Brinkmann
Alpha Heating Circulators, Lauda-Brinkmann
Alpha Heating Circulators, Lauda-Brinkmann
Alpha Heating Circulators, Lauda-Brinkmann
The LAUDA Alpha heating and cooling circulators provide a temperature range from -25 to 100 °C, depending on the exact model
The Alpha heating and cooling circulators are suitable for operation with non-flammable liquids (water, water/glycol) and for both internal and external temperature control tasks. A one point calibration of the thermostats can be carried out by the user. Stainless steel bath.
This temperature range was selected with a focus of addressing most of the common temperature control applications within the laboratory.   This entry level equipment range is made possible by minimizing any unnecessary features, with the focus on reliability and user-friendliness.
  • Catalog No:
  • 97039-882
  • 97039-884
  • 97039-886
  • Pump flow rate:
  • 15 L/min. (Max.)
  • 15 L/min. (Max.)
  • 15 L/min. (Max.)
  • Pump pressure:
  • .2 Bar (Max.)
  • .2 Bar (Max.)
  • .2 Bar (Max.)
Product Family Options
  • Modèle
    Plage de température
    Electrical
    Puissance cryo
    Heating Power
  • A 6
    25...100 °C
    115 V, 60 Hz
    0
    1.15kW
    Each
    Each - $7,523.22
    $7,523.22
    Each
       $7,523.22
     : L000631
    Specifications
    Cat. No.
    97039-882
    Pump flow rate
    15 L/min. (Max.)
    Pump pressure
    .2 Bar (Max.)
  • A 12
    25...100 °C
    115 V, 60 Hz
    0
    1.15kW
     : -
    Specifications
    Cat. No.
    97039-884
    Pump flow rate
    15 L/min. (Max.)
    Pump pressure
    .2 Bar (Max.)
  • A 24
    25...100 °C
    115 V, 60 Hz
    0
    1.15kW
    Each
    Each - $9,790.26
    $9,790.26
    Each
       $9,790.26
     : L000633
    Specifications
    Cat. No.
    97039-886
    Pump flow rate
    15 L/min. (Max.)
    Pump pressure
    .2 Bar (Max.)