Five-legged stool offers increased stability and helps to prevent students or employees from leaning back on two legs, similar to a five-legged office chair
14" x 1-1/4" thick round high pressure laminate with virtually indestructible Lotz Armor Edge seat.
14" round seat is constructed of medium density fiberboard core with threaded inserts for mounting seat to fully welded black textured frame. Virtually indestructible Lotz Armor edge is a spray-on application like a truck bed liner in black. Seat top is high pressure laminate in choice of bannister oak or grey nebula. Black non-marring glides. Frame is constructed of 16-gauge steel tube, 1.25" for adjustable and 1" for fixed legs with fully welded 12-gauge seat mounting plate and .375" round foot rest ring.
Fully-welded steel stools are durable and withstand rigorous classroom, lab or plant use for years to come. Five-legged stools are constructed of 16-gauge, heavy-duty powder-coated steel tube legs welded to a 12-gauge steel seat plate and .375" round foot ring welded to the frame. 14" x 1.25" high pressure laminate with indestructible Lotz Armor Edge banding. Non-marring glides.
- Catalog No:
- 10054-706
- 10054-704
- 10054-710
- 10054-708
- 10054-906
- 10054-910
- 10054-908
- 10054-912
- Model:
- 10054-706
- 10054-704
- 10054-710
- 10054-708
- 10054-906
- 10054-910
- 10054-908
- 10054-912
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified