VWR® Petri Dishes with Markings
Catalogue # CA73380-016
Fournisseur: VWR International
Spécifications
- Description:Semi-Stacking Petri Dish with 6 Vents
- Dimensions:150 x 15 mm
- Cat. no.:CA73380-016
Spécifications
À propos de cet article
These Petri dishes are ideal for antibiotic susceptibility testing.
- Accommodate two to three membrane filters
- Suitable for bioburden determination and sterility testing
- Extremely flat bottom
They accommodate two to three membrane filters per dish, and they are large enough for bioburden determination and sterility testing of medical devices. The extremely flat bottom has markings for placing up to six strips of Etest®.