Pasteur Pipettes, Argos Technologies | Avantor
Pasteur Pipettes, Argos Technologies
Pasteur Pipettes, Argos Technologies
Supplier:  Antyila Scientific
Certificates
Pasteur Pipettes, Argos Technologies
Pasteur Pipettes, Argos Technologies
2 Options Available Below
Product Details & Documents
Argos plastic pasteur pipettes are produced from medical-grade polystyrene in a class 100,000 cleanroom.
Pipettes are available in two sizes and are supplied individually wrapped to maintain sterility. They are gamma irradiated, DNase/Rnase-free, and non pyrogenic.
Plastic pasteur pipettes offer the aspirating precision of glass, and the safety and benefits of plastic. Plastic construction allows the long flexible stem to be bent to draw liquid from narrow or small volume tubes. These pipettes can be used with aspirating bulbs or vaccum applications to remove liquid waste from flasks, dishes, plates, and tubes.
Product Family Options
Informations sur le produit
  • Désignation
    Overall Length
    Sterility
    Packed
    Availability
    Price
  • Polystyrene Pasteur Pipette
    146 mm (5.75")
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 200
    Case of 200 - $229.26
    $229.26
    Prix promotionnel pour les nouveaux clients
    Case of 200
       $229.26
    Supplier Number: 04395-11
  • Polystyrene Pasteur Pipette
    229 mm (9")
    Sterile
    Individually Wrapped
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 200
    Case of 200 - $198.56
    $198.56
    Prix promotionnel pour les nouveaux clients
    Case of 200
       $198.56
    Supplier Number: 04395-12