Batch Ball and Rod Mills, Sepor
Fournisseur: SEPOR CA
Certificats
À propos de cet article
Designed to be companion equipment to the SEPOR drive rolls, these durable containers will grind from 1000 to 2500 g charges.
- In the stainless steel models, all wetted parts are stainless steel
- Grinding media is also available
Heavy-duty construction, including machined rolling rings, a quick-threaded removable yoke, a rubber gasketed lid with handles, and interior lifter bars. Both ball and rod mills are available in mild steel, 1/4" urethane-lined mild steel, and stainless steel constructions.
Informations sur les accessoires: The charge for ball mills is 20 lbs. which is composed of 1.25" and 1" hardened alloy steel balls. For the rod mill, a set of round steel rods 3.4" (9 each), 5/8" (8 each), 1/2" (7 each), by 9" long. Stainless steel balls are mixed sizes and rods are also standard charges.
Ordering information: Contact your Avantor sales representative for more information.
Frequently Bought Together