ContactUsLinkComponent
Wet-Dry Sieve Shaker, Sepor
Wet-Dry Sieve Shaker, Sepor
Catalog # CA11020-358
Supplier:  SEPOR CA
Wet-Dry Sieve Shaker, Sepor
Catalog # CA11020-358
Supplier:  SEPOR CA
Supplier Number:  040B-001
Specifications
  • Description:
    Wet-Dry Sieve Shaker, 110V/60Hz
  • Cat. no.:
    CA11020-358
  • Supplier No.:
    040B-001
Specifications
About this item
The Wet-Dry Sieve Shaker is supplied for the purpose of conducting relatively simple size analysis for materials
The sieve shaker uses standard 8 inch diameter laboratory sieve screens and has a capacity for 2 full height 8" diameter sieve screens. The smallest opening sieve screen that should be used with the Wet-Dry Sieve Shaker is 100 mesh.
Ordering information: Contact your Avantor sales representative for more information.