Cupel Tray and Handle, Legend
Supplier: Legend
Certificates
About this item
Cupel trays are not made to put into the furnace during the cupellation process
Rather, they are used for keeping the cupels in proper order and location before, and after the cupellation process. Hot cupels can be placed onto the tray after removal from the oven. Also used for keeping lead buttons in order prior to cupellation. For use with cupels up to 13/4" diameter.
Ordering information: Contact your Avantor sales representative for more information.
Frequently Bought Together